Chromium(II) chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Chromium dichloride

M-Clarity™ quality level = MQ200

8.02482.0010
EM8.02482.0010CS 15354 USD
EM8.02482.0010
Chromium(II) chloride for synthesis, Sigma-Aldrich®
Chromium (II) chloride
Formula: Cl₂Cr
Boiling Pt: 1302 °C (1013 hPa)
Melting Pt: 823…825 °C
Density: 2.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8,III
Merck Index: 13,02264

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 823 - 825 °C
Molar Mass 122.9 g/mol
Boiling Point 1300 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR