4-tert-Butylpyrocatechol, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01987.1000
EM8.01987.1000EA 240 USD
EM8.01987.1000
4-tert-Butylpyrocatechol, Sigma-Aldrich®
4-tert-Butylpyrocatechol
Formula: C₁₀H₁₄O₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.05 g/cm³ (60 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00002201
CAS Number: 98-29-3
EINECS: 202-653-9
UN: 2923
ADR: 8 (6.1),III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR