Benzophenone, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Diphenylmethanone, Diphenyl ketone
M-Clarity™ quality level = MQ200
8.01801.0250
8.01801.1000
EM8.01801.0250EA
33
USD
EM8.01801.0250
EM8.01801.1000
Benzophenone, Sigma-Aldrich®
Benzophenone
Formula:
C₁₃H₁₀O Boiling Pt: 305.4 °C (1013 hPa) Melting Pt: 47…49 °C Density: 1.085 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 EINECS: 204-337-6 UN: 3077 ADR: 9,III Merck Index: 13,01099 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...