1-Chlorobutane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.01640.0100
8.01640.1000
8.01640.2500
EM8.01640.0100CS
593
USD
EM8.01640.0100
EM8.01640.1000
EM8.01640.2500
1-Chlorobutane for synthesis, Sigma-Aldrich®
1-Chlorobutane
Formula:
CH₃(CH₂)₃Cl Boiling Pt: 78.4 °C (1013 hPa) Melting Pt: –123 °C Density: 0.8862 g/cm³ (20 °C) Flash Pt: –17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001009 CAS Number: 109-69-3 EINECS: 203-696-6 UN: 1127 ADR: 3,II Merck Index: 13,01559 |
|
Specification Test Results
Solubility | 0.5 g/l (20 °C) |
Melting Point | -123 °C |
Molar Mass | 92.57 g/mol |
Boiling Point | 79 °C (1013 hPa) |
Vapor Pressure | 110 hPa (20 °C) |
Flash Point | -17 °C |
Explosion Limit | 1.8 - 10.1 %(V) |
Density | 0.886 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 280 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...