1,4-Butanediol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.01532.2500 8.01532.9025
EM8.01532.2500CS 406 USD
EM8.01532.2500 EM8.01532.9025
1,4-Butanediol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
1,4-Butanediol
Formula: C₄H₁₀O₂
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 19.5…20.0 °C
Density: 1.02 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002968
CAS Number: 110-63-4
EINECS: 203-786-5

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 19.5 - 20.0 °C
Molar Mass 90.12 g/mol
Boiling Point 230 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 134 °C
Refractive Index 1.4446 (25 °C, 589 nm)
Explosion Limit 2.4 - 15.3 %(V)
Density 1.02 g/cm3 (20 °C)
pH 7 - 8 (500 g/l, H2O, 20 °C)
Ignition Point 385 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR