4-Aminopyridine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Pyridin-4-amine, 4-Pyridylamine

M-Clarity™ quality level = MQ200

8.01111.0010 8.01111.0025
EM8.01111.0010CS 5423 USD
EM8.01111.0010 EM8.01111.0025
4-Aminopyridine for synthesis, Sigma-Aldrich®
4-Aminopyridine
Formula: C₅H₆N₂
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 157…160 °C
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00006439
CAS Number: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II
Merck Index: 13,03964

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 157 - 160 °C
Molar Mass 94.11 g/mol
Boiling Point 274 °C (1013 hPa)
Flash Point 156 °C
Density 1.26 g/cm3 (25 °C)
Ignition Point 640 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR