Diethyl malonate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester
M-Clarity™ quality level = MQ200
8.00898.0100
8.00898.0500
8.00898.1000
8.00898.2500
EM8.00898.0100CS
748
USD
EM8.00898.0100
EM8.00898.0500
EM8.00898.1000
EM8.00898.2500
Diethyl malonate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Diethyl malonate
Formula:
C₇H₁₂O₄ MW: 160.17 g/mol Boiling Pt: 199 Melting Pt: -50 Density: 1.055 g/cm³ (20 °C) Flash Pt: 93 Storage Temperature: Ambient |
MDL Number:
MFCD00009195 CAS Number: 105-53-3 EINECS: 203-305-9 Merck Index: 13,03856 |
Specification Test Results
Assay (GC, area%) | ≥ 98.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 1.053 - 1.057 |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...