Iodoethane ≥98.0% (by GC) stabilized for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Stabilization:
Stabilised with silver
M-Clarity™ quality level = MQ200
8.00896.0100
8.00896.0500
8.00896.9050
EM8.00896.0100CS
2404
USD
EM8.00896.0100
EM8.00896.0500
EM8.00896.9050
Iodoethane ≥98.0% (by GC) stabilized for synthesis, Sigma-Aldrich®
Iodoethane
Formula:
C₂H₅I Boiling Pt: 69…73 °C (1013 hPa) Melting Pt: –108 °C Density: 1.93 g/cm3 (20 °C) Flash Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001091 CAS Number: 75-03-6 EINECS: 200-833-1 UN: 1993 ADR: 3,III Merck Index: 13,03846 |
Specification Test Results
Solubility | 4 g/l (20 °C) (slow decomposition) |
Melting Point | -108 °C |
Molar Mass | 155.97 g/mol |
Boiling Point | 69 - 73 °C (1013 hPa) |
Vapor Pressure | 133 hPa (18 °C) |
Flash Point | 53 °C |
Refractive Index | 1.513 (20 °C, 589 nm) |
Density | 1.93 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...