Iodoethane ≥98.0% (by GC) stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


Stabilization: Stabilised with silver
M-Clarity™ quality level = MQ200

8.00896.0100 8.00896.0500 8.00896.9050
EM8.00896.0100CS 2404 USD
EM8.00896.0100 EM8.00896.0500 EM8.00896.9050
Iodoethane ≥98.0% (by GC) stabilized for synthesis, Sigma-Aldrich®
Iodoethane
Formula: C₂H₅I
Boiling Pt: 69…73 °C (1013 hPa)
Melting Pt: –108 °C
Density: 1.93 g/cm3 (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001091
CAS Number: 75-03-6
EINECS: 200-833-1
UN: 1993
ADR: 3,III
Merck Index: 13,03846

Order Now

Specification Test Results

Solubility 4 g/l (20 °C) (slow decomposition)
Melting Point -108 °C
Molar Mass 155.97 g/mol
Boiling Point 69 - 73 °C (1013 hPa)
Vapor Pressure 133 hPa (18 °C)
Flash Point 53 °C
Refractive Index 1.513 (20 °C, 589 nm)
Density 1.93 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR