2-Ethoxyethanol ≥99.9% (by GC) stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00857.1000 8.00857.2500
EM8.00857.1000CS 372 USD
EM8.00857.1000 EM8.00857.2500
2-Ethoxyethanol ≥99.9% (by GC) stabilized for synthesis, Sigma-Aldrich®
2-Ethoxyethanol
Formula: C₂H₅OCH₂CH₂OH
Boiling Pt: 135 °C (lit.)
Melting Pt: –90 °C (lit.)
Density: 0.93 g/ml (25°C) (lit.)
Flash Pt: 40 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
REACH: 01-2119560582-38
Merck Index: 13,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -100 °C
Molar Mass 90.12 g/mol
Boiling Point 135 °C (1013 hPa)
Vapor Pressure 5 hPa (20 °C)
Flash Point 40 °C
Refractive Index 1.408 (20 °C, 589 nm)
Explosion Limit 1.8 - 15.7 %(V)
Density 0.93 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 235 °C
Saturation concentration 18 g/m3 (20 °C) Air
Evaporation number 43
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR