Diethyl sebacate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00757.0250
EM8.00757.0250CS 1180 USD
EM8.00757.0250
Diethyl sebacate for synthesis, Sigma-Aldrich®
Diethyl sebacate
Formula: C₁₄H₂₆O₄
MDL Number: MFCD00009219
CAS Number: 110-40-7

Order Now

Specification Test Results

Solubility 0.08 g/l (20 °C)
Melting Point -3 °C
Molar Mass 258.35 g/mol
Boiling Point 312 °C (1013 hPa)
Flash Point 71 °C
Density 0.97 g/cm3 (20 °C)
Viscosity kinematic 6.2 mm2/s (20 °C)
Synonyms Sebacic acid diethyl ester, Decanedioic acid diethyl ester
Hill Formula C14H26O4
Chemical Formula C2H5OOC(CH2)8COOC2H5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR