Sebacic acid ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00753.9050
EM8.00753.9050EA 1365 USD
EM8.00753.9050
Sebacic acid ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Sebacic acid
Formula: C₁₀H₁₈O₄
Boiling Pt: 295 °C (133 hPa)
Melting Pt: 130…133 °C
Density: 1.21 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6

Order Now

Specification Test Results

Solubility 1 g/l (20°C)
Melting Point 130 - 133 °C
Molar Mass 600 - 620 kg/m3
Bulk Density 600 - 620 kg/m3
Boiling Point 295 °C (133 hPa)
Vapor Pressure 1.3 hPa (183 °C)
Density 1.21 g/cm3
Synonyms Decanedioic acid
Hill Formula C10H18O4
Chemical Formula HOOC(CH2)8COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR