Selenium dioxide for synthesis, sublimated, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00653.0005
EM8.00653.0005EA 46 USD
EM8.00653.0005
Selenium dioxide for synthesis, sublimated, Sigma-Aldrich®
Selenium dioxide
Formula: SeO₂
MDL Number: MFCD00003562
CAS Number: 7446-08-4
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 384 g/l (14°C)
Molar Mass 1500 kg/m3
Bulk Density 1500 kg/m3
Vapor Pressure 16 hPa (70 °C)
pH 2 (10 g/l, H2O, 20 °C)
Synonyms Selenious anhydride, Selenium(IV) oxide
Hill Formula O2Se
Chemical Formula SeO2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR