Propionic acid, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Propanoic acid

M-Clarity™ quality level = MQ200

8.00605.9025
EM8.00605.9025EA 584 USD
EM8.00605.9025
Propionic acid, Sigma-Aldrich®
Propionic acid
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: –21 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00002756
CAS Number: 79-09-4
UN: 3463
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR