Methacrylic acid, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Methylpropenoic acid

M-Clarity™ quality level = MQ200

8.00578.9025
EM8.00578.9025EA 829 USD
EM8.00578.9025
Methacrylic acid, Sigma-Aldrich®
Methacrylic acid
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163
Melting Pt: 15
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 68
MDL Number: MFCD00002651
CAS Number: 79-41-4
UN: 2531
ADR: 8 II
Merck Index: 13,05967

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR