Butyric acid ≥99% (by GC), Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00457.9050
EM8.00457.9050EA 1117 USD
EM8.00457.9050
Butyric acid ≥99% (by GC), Sigma-Aldrich®
Butyric acid
Formula: C₄H₈O₂
Boiling Pt: 162…165 °C (1013 mbar)
Melting Pt: –8…–5 °C
Density: 0.96 g/cm3 (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
UN: 2820
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR