4-Aminophenol ≥99.0% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.00421.0005 8.00421.0250 8.00421.1000
EM8.00421.0005CS 35361 USD
EM8.00421.0005 EM8.00421.0250 EM8.00421.1000
4-Aminophenol ≥99.0% for synthesis, Sigma-Aldrich®
4-Aminophenol
Formula: H₂NC₆H₄OH
Boiling Pt: 284 °C (1013 hPa) (dec.)
Melting Pt: 190 °C
Density: 1.3 g/cm³ (20 °C)
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00007869
CAS Number: 123-30-8
EINECS: 204-616-2
UN: 2512
ADR: 6.1,III
Merck Index: 13,00461
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7af17b754d4ebaa0993b4544d4e6ab6c116724c&vid=6275667c2624ee686f61715acd420dc596e076fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (HClO₄) ≥ 99.0 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR