4-Aminobutyric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: GABA, γ-Aminobutyric acid

M-Clarity™ quality level = MQ200

8.00302.0100 8.00302.9025
EM8.00302.0100CS 990 USD
EM8.00302.0100 EM8.00302.9025
4-Aminobutyric acid for synthesis, Sigma-Aldrich®
4-Aminobutyric acid
Formula: C₄H₉NO₂
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 200…202 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 200 - 202 °C
Molar Mass 103.12 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR