Caprylic acid, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

8.00192.1000
EM8.00192.1000EA 168 USD
EM8.00192.1000
Caprylic acid, Sigma-Aldrich®
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7
Melting Pt: 16
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8 II
Merck Index: 13,01771

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR