Barbituric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00133.0100 8.00133.0250
EM8.00133.0100CS 1139 USD
EM8.00133.0100 EM8.00133.0250
Barbituric acid for synthesis, Sigma-Aldrich®
Barbituric acid
Formula: C₄H₄N₂O₃
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00006666
CAS Number: 67-52-7
EINECS: 200-658-0
Merck Index: 13,00966

Order Now

Specification Test Results

Solubility 11.45 g/l (25 °C)
Melting Point 250 - 252 °C (decomposition)
Molar Mass 128.08 g/mol
Bulk Density 720 kg/m3
pH 2 - 3 (50 g/l, H2O, 60 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR