Benzonase® Nuclease
Supplier: MilliporeSigma
71205-3
EM71205-3EA 412.21 USD
EM71205-3
Benzonase® Nuclease
Enzymes
Effective viscosity reduction and removal of nucleic acids from protein solutions.
  • The 99% purity grade is tested for endotoxins and contains 0.25 EU/1000 units
  • It is supplied as a 0.2 µm filtered solution in 50% glycerol
  • These are genetically engineered endonuclease from Serratia marcescens
  • Stores at –20 °C
Benzonase Nuclease is available in ultrapure and pure grades at a standard concentration of 25-29 U/µL and at a high concentration (HC) of 250 U/µL. Both preparations are free of detectable protease and have specific activity 1 × 10⁶ U/mg protein. It degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) while having no proteolytic activity. It is effective over a wide range of conditions and possesses an exceptionally high specific activity.