dNTP Mix, 10mM, MilliporeSigma
Supplier: MilliporeSigma
71004-3
EM71004-3EA 115.67 USD
EM71004-3
dNTP Mix, 10mM, MilliporeSigma
Nucleic Acid Reagents Nucleotides
The dNTP Mix contains ultrapure dATP, dCTP, dGTP, and dTTP (monosodium salts) at a concentration of 10 mM each in sterile deionized water at pH 7.0.
  • Ready to use
  • RNase and DNase free
Ideal for any application that requires pure deoxynucleotides, such as PCR, cDNA synthesis, and fill-in reactions.