SpinPrep™ Gel DNA Extraction Kit, MilliporeSigma
Supplier: MilliporeSigma
70852-3
EM70852-3EA 419.54 USD
EM70852-3
SpinPrep™ Gel DNA Extraction Kit, MilliporeSigma
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The SpinPrep™ Gel DNA Kit enables efficient extraction of DNA fragments of 150 bp to > 12,000 bp in size from agarose gels.
  • No organic extraction or alcohol precipitation
  • Total preparation time is less than 30 minutes
  • No low melting point agarose is required
The procedure uses GelMelt™ Solution to dissolve the gel slice, followed by adsorption of the DNA to a silica membrane in a spin column format. After a wash step, the purified DNA is eluted in low-salt buffer. Each spin column can bind up to 20 µg DNA. Recoveries are consistently in the range of 50-90% under standard conditions.