4X SDS Sample Buffer, MilliporeSigma
Supplier: MilliporeSigma
70607-3
EM70607-3EA 65.06 USD
EM70607-3
4X SDS Sample Buffer, MilliporeSigma
Buffers Electrophoresis Buffers Loading Buffers
The 4X SDS Sample Buffer is a standard formulation commonly used for SDS-PAGE analysis of proteins
The solution includes DTT for complete denaturation of disulfide bonds. The buffer can be used at 2X for most applications.