Novagen® GST Bind™ Resin, MilliporeSigma
Supplier: MilliporeSigma
70541-3 70541-4 70541-5
EM70541-3EA 448.87 USD
EM70541-3 EM70541-4 EM70541-5
Novagen® GST Bind™ Resin, MilliporeSigma
Protein Purification Protein Purification Resins
GST Bind™ Resin utilizes an 14-atom spacer arm to covalently attach reduced glutathione via a sulfide linkage. The high degree of substitution of glutathione ensures a high binding capacity with yields of glutathione S-transferase (GST) fusion proteins of 5 to 8 mg/ml settled resin. The resin can be re-used up to 6 times without loss of capacity.