Novagen® pET-28b(+) DNA, MilliporeSigma
Supplier: MilliporeSigma
69865-3
EM69865-3EA 477.33 USD
EM69865-3
Novagen® pET-28b(+) DNA, MilliporeSigma
Vectors, Plasmids and Libraries
The pET-28a-c(+) vectors carry an N-terminal His Tag®/thrombin/T7 Tag® configuration plus an optional C-terminal His Tag sequence.
  • Sequence is numbered by the pBR322 convention
  • T7 expression region is reversed on the vector map (TB074)
The f1 origin is oriented so that infection with helper phage will produce virions containing single-stranded DNA that corresponds to the coding strand. The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock).