Novagen® pET-28a(+) DNA, MilliporeSigma
Supplier: MilliporeSigma
69864-3
EM69864-3EA
484.04
USD
EM69864-3
Novagen® pET-28a(+) DNA, MilliporeSigma
Vectors, Plasmids and Libraries
The pET-28a-c(+) vectors carry an N-terminal His Tag/thrombin/T7 Tag configuration plus an optional C-terminal His Tag sequence.
- Sequence is numbered by the pBR322 convention
- T7 expression region is reversed on the vector map (TB074)
The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock).