Novagen® pET-21a Vectors, MilliporeSigma
Supplier: MilliporeSigma
69740-3
EM69740-3EA
475.06
USD
EM69740-3
Novagen® pET-21a Vectors, MilliporeSigma
Vectors, Plasmids and Libraries
The pET-21a vectors carry an N-terminal T7 Tag® sequence plus an optional C-terminal His Tag® sequence.
- Vectors differ from pET-24a only by their selectable marker
- Sequence is numbered by the pBR322 convention
- T7 expression region is reversed on the vector map (TB036)
The f1 origin is oriented so that infection with helper phage will produce virions containing single-stranded DNA that corresponds to the coding strand.The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock).It is shipped with blue ice or dry ice.