Anti-IgG Goat Antibody (AP (Alkaline Phosphatase))

Supplier: MilliporeSigma
69265-3
EM69265-3EA 123.58 USD
EM69265-3
Anti-IgG Goat Antibody (AP (Alkaline Phosphatase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR