Genomic DNA, Mouse, EMD Millipore
Supplier: MilliporeSigma
69239-3
EM69239-3EA 363.07 USD
EM69239-3
Genomic DNA, Mouse, EMD Millipore
Electrophoresis Reagents Western Blotting Reagents
Genomic DNAs are highly purified preparations of high molecular weight genomic DNA from a variety of organisms.
The isolation protocol minimizes shearing and removes contaminants that inhibit enzymatic procedures. Novagen genomic DNA is qualified for genomic analysis (including PCR) and library construction. At least 90% of the DNA is greater than 100 kbp in size, as analyzed by CHEF gel electrophoresis (> 50 kbp for S. cerevisiae). No inhibition of restriction enzyme activity is observed, and the DNA is free of contaminating RNA and protein.
Delivery information: DNA is supplied in 10 mM Tris-HCl, pipet with a wide-bore pipet tip to minimize shearing.