Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥94% (by HPLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ100

573115-50MG
EM573115-50MGEA 212.49 USD
EM573115-50MG
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥94% (by HPLC), Calbiochem®, Millipore®
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

SMCC, Water-Soluble, a water-soluble derivative of SMCC.


Ideal for coupling enzymes to antibodies. Contains a cyclohexane bridge, which gives extra stability to the maleimide group. Both enzyme activity and antibody specificity are preserved after coupling. Water-soluble cross-linking reagents help to avoid the use of organic solvents such as dimethylformamide (DMF) and dimethylsulfoxide (DMSO) when preparing bioconjugates.


Ideal for coupling enzymes to antibodies. Contains a cyclohexane bridge, which gives extra stability to the maleimide group. Both enzyme activity and antibody specificity are preserved after coupling.

Formula: C₁₆H₁₈N₂O₉S
Storage Temperature: Freezer
MDL Number: MFCD00054978
CAS Number: 92921-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR