N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate (SMCC) ≥97% (by HPLC), Millipore®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ100

573114-100MG
EM573114-100MGEA 269.36 USD
EM573114-100MG
N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate (SMCC) ≥97% (by HPLC), Millipore®
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

Heterobifunctional reagent useful for enzyme labeling of antibodies and antibody fragments. Cross-links both a sulfhydryl group and a free amine group. Useful for sequential conjugation to minimize polymerization.
Reacts with primary amines and sulfhydryl groups.

Formula: C₁₆H₁₈N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00009634
CAS Number: 64987-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR