Aquatic Block™ Reagent, Salmon Plasma
Supplier: MilliporeSigma
558300-500ML
EM558300-500MLEA 230.02 USD
EM558300-500ML
Aquatic Block™ Reagent, Salmon Plasma
Plasma
A ready-to-use blocking reagent for Western blotting and immunoassays. Contains salmon plasma proteins that do not interact with mammalian antibodies, thus offering a reduced background.