Protease Assay Kit, EMD Millipore
Supplier: MilliporeSigma
539125-1KIT
EM539125-1KITEA 1146.66 USD
EM539125-1KIT
Protease Assay Kit, EMD Millipore
Assays Protein Assays
This kit suitable for quantitative array of a wide variety of proteases in biological samples, utilizing FTC-casein as a substrate.
  • Protect from light
  • SUitable for colorimetric detection method
Used to detect the enzyme activities of chymotrypsin, elastase, plasminogen, PRONASE® Protease, subtilisin, thermolysin and trypsin.
Delivery information: Kit contains fluorescent-casein substrate, incubation buffer, assay buffer trypsin standard, and a user protocol.
Caution: FTC-casein, protease positive control, incubation buffer should be stored at –20 °C.