MOWIOL® 4-88 Reagent, Gelvatol, Millipore®

Supplier: MilliporeSigma

475904-100GM
EM475904-100GMEA 187.3 USD
EM475904-100GM
MOWIOL® 4-88 Reagent, Gelvatol, Millipore®
Polyvinyl alcohol

Yellow solid used for mounting cells on slides for detection of cell-surface antigens by immunofluorescence


  • pH 4.5 - 7.0


Useful as an anti-fading agent.

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR