Calbiochem® Miracloth, EMD Millipore
Supplier: MilliporeSigma
475855-1R
EM475855-1REA 192.56 USD
EM475855-1R
Calbiochem® Miracloth, EMD Millipore
Filters Membrane Filters
Miracloth is an autoclavable, quick filtration material for gelatinous homogenates that is composed of rayon-polyester with an acrylic binder.
It is ideal for filtration of cleared neutralized lysates during medium- and large-scale plasmid purification to prevent contamination with loosely adherent pellet material and is useful for protoplast isolation.