Anti-PhosphoDetect Insulin Receptor (phospho-Tyr1162/1163) Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
407707-10T
EM407707-10TEA 636.49 USD
EM407707-10T
Anti-PhosphoDetect Insulin Receptor (phospho-Tyr1162/1163) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PhosphoDetect Insulin Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR