Anti-IP₃ Receptor Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
407143-100UG
EM407143-100UGEA 560.4 USD
EM407143-100UG
Anti-IP₃ Receptor Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody purified by ammonium sulfate precipitation and ion-exchange (DEAE) column chromatography. Recognizes the ~260 kDa IP₃ receptor protein.

Type: Primary
Antigen: IP₃ Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Dog;Mouse;Rat;Pig;Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR