Anti-Glucose Transporter-4 Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
400064-50UG
EM400064-50UGEA 483.06 USD
EM400064-50UG
Anti-Glucose Transporter-4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Glucose Transporter-4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR