Anti-GLUT2 Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
400061-50UG
EM400061-50UGEA 517.09 USD
EM400061-50UG
Anti-GLUT2 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Glucose Transporter-2 Reacts with Human, Mouse, Rat

Type: Primary
Antigen: Glucose Transporter-2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR