Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv® for liquid chromatography, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

4.80448.2500
EM4.80448.2500CS 1953 USD
EM4.80448.2500
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv® for liquid chromatography, Supelco®
Acetonitrile with 0.1% (v/v) Trifluoroacetic acid

Offering a high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. They are produced from specially selected raw materials, and undergo a number of purification steps prior to final packaging. Since separations are normally carried out under gradient conditions in analytical HPLC, Merck Millipore offer solvents in ’gradient grade’ as well as ’isocratic grade’. This enables you to minimise the gradient effect of the solvent involved.


Acetonitrile with 0.1% (v:v) trifluoroacetic acid for liquid chromatography LiChrosolv®

Density: 0.786 g/cm3 (20 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
EINECS: 200-929-3
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Assay (trifluoroacetic acid) 0.095 - 0.105 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR