Trifluoroacetic acid 0.1% (v/v) in water, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

4.80112.2500
EM4.80112.2500CS 1143 USD
EM4.80112.2500
Trifluoroacetic acid 0.1% (v/v) in water, Supelco®
Trifluoroacetic acid

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.

Formula: C₂HF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR