Anti-HNE Fluorophore Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
393206-100UL
EM393206-100ULEA 538.61 USD
EM393206-100UL
Anti-HNE Fluorophore Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-HNE Fluorophore Reacts with Pan

Type: Primary
Antigen: HNE Fluorophore
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR