Helix pomatia Native Beta-glucuronidase
Supplier: MilliporeSigma
347420-1MU
EM347420-1MUEA 444.63 USD
EM347420-1MU
Helix pomatia Native Beta-glucuronidase
Enzymes
Useful for hydrolyzing urinary steroid conjugates prior to steriod assays.