Anti-Ganglioside Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
345759-100UL
EM345759-100ULEA 1158.53 USD
EM345759-100UL
Anti-Ganglioside Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Ganglioside Reacts with Pan

Type: Primary
Antigen: Ganglioside
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR