Anti-Ganglioside GM₁ Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
345757-100UL
EM345757-100ULEA 538.61 USD
EM345757-100UL
Anti-Ganglioside GM₁ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Ganglioside GM₁
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR