Anti-Factor B Goat Polyclonal Antibody

Supplier: MilliporeSigma
341272-2ML
EM341272-2MLEA 249.2 USD
EM341272-2ML
Anti-Factor B Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Factor B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR