EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98%, Calbiochem®

Supplier: MilliporeSigma
Warning


Packaged under inert gas.

341006-25GM 341006-5GM
EM341006-25GMEA 490.7 USD
EM341006-25GM EM341006-5GM
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98%, Calbiochem®
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

EDAC HCl is a water-soluble derivative of carbodiimide useful for conjugating haptens to proteins and polypeptides.


Used to modify NMDA receptors and as a condensing agent in peptide synthesis.


Special Instructions: Unstable in solution. Reconstitute just prior to use.

Formula: C₈H₁₇N₃·HCl
Melting Pt: 112…113 °C
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% by Titration
Form White solid
Molecular Formula C₈H₁₇N₃ · HCl
Molecular Weight 191.7
Toxicity Irritant
Storage -20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR