Ethidium Bromide Adsorber
Supplier: MilliporeSigma
Gray-brown suspension
- CAS No.: 69011-20-7
- Storage +15 °C to +30 °C
Developed specifically for the safe and simple removal of ethidium bromide (EtBr) from aqueous staining solutions and running buffers used in nucleic acid separation gels.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...