Anti-PhosphoDetect EGFR (phospho-Tyr1068) Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
324867-10T
EM324867-10TEA 649.94 USD
EM324867-10T
Anti-PhosphoDetect EGFR (phospho-Tyr1068) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PhosphoDetect EGFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Chicken;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR